Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Camphor Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019631 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9631 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9631 Supplier MilliporeSigma Supelco Supplier No. PHR11191G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. D-Camphor is an acyclic terpenoid and one of the key volatile constituents of essential oils of plants Artemisia tridenta, Mentha longifolia ssp., Longifolia and some Lamiaceae species. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 464-49-3
Ignition Point 870°F
Vapor Pressure 4 mmHg (70°C)
Vapor Density 5.24 (vs. air)
Flash Point 66°C
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C10H16 O
MDL Number MFCD00064149
Melting Point 178°C to 182°C
Synonym 2-Bornanone; 2-Camphanone
Type Certified Reference Material
Quantity 1 g
Formula Weight 152.23
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.