Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Camphoric acid, 99%

Catalog No. AC108205000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108205000 500 g Plastic bottle
AC108200050 5 g Glass bottle
AC108201000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC108205000 Supplier Thermo Scientific Chemicals Supplier No. 108205000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 124-83-4 | C10H14O4-2 | 198.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-83-4
Molecular Formula C10H14O4-2
Molecular Weight (g/mol) 198.218
MDL Number MFCD00001375
InChI Key LSPHULWDVZXLIL-LDWIPMOCSA-L
Synonym d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
PubChem CID 6918944
IUPAC Name (1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
SMILES CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C

Specifications

Melting Point 183°C to 187°C
Color White
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Assay Percent Range 99%
Beilstein 09, 745
Fieser 11,352; 15,67
Merck Index 15, 1735
Formula Weight 200.23
Percent Purity ≥98.5%
Chemical Name or Material D(+)-Camphoric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.