Learn More
Medchemexpress LLC D-Carnitine | 541-14-0 | 99.9% | 161.20 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW012550
Encompass_Preferred
D-Carnitine is an orally available isomer of the essential nutrient L-carnitine that promotes long-chain fatty acid transport into the mitochondrial matrix for beta-oxidation. It also exhibits antiparasitic activity.
  • Orally available isomer of L-carnitine.
  • Promotes long-chain fatty acid transport into the mitochondrial matrix for beta-oxidation.
  • Has antiparasitic activity.
  • Inhibits the uptake of L-carnitine by renal brush marginal membrane vesicles in rats (at 0.5 μM).
  • Inhibits the uptake of L-carnitine by primary human limbal (HCLE) and conjunctival epithelium (HCjE) cells (at 0.5, 1 μM).
  • Inhibits parasite activity in rats and mice infected with trypanosoma (at 75-300 mg/kg/day, orally).
Catalog No. 50-004-51407