Learn More
Cayman Chemical D-Carnitine, Purity: ≥98%, CAS: 541-14-0, 50 mg
SDP
Catalog No. 502980596
Encompass_Preferred
An isomer of L-carnitine; inhibits L-carnitine uptake by rat kidney brush border membrane vesicles at 50 µM and by primary HCLE and HCjE cells at 0.5 and 1 mM
Catalog No. 50-298-0596 Supplier Cayman Chemical Supplier No. 1674950 MG
An isomer of L-carnitine; inhibits L-carnitine uptake by rat kidney brush border membrane vesicles at 50 µM and by primary HCLE and HCjE cells at 0.5 and 1 mM