Learn More
Alkali Scientific D (+) Cellobiose, 25 Grams
SDP
Catalog No. 5000660231
Encompass_Preferred
D (+) Cellobiose is a high-purity carbohydrate used in laboratory applications. This 25-gram quantity is ideal for experiments requiring a precise amount of cellobiose, a disaccharide composed of two glucose molecules. It is often used in biochemical research, especially in the study of enzymes, fermentation processes, and metabolic pathways involving cellobiose as a substrate. The product is of pharmaceutical grade, ensuring high quality and purity for accurate results in laboratory experiments. Its use extends to microbiology, molecular biology, and carbohydrate analysis protocols. Available in a convenient 25g bottle, perfect for routine testing and experiments.
Catalog No. 50-006-60231 Supplier Alkali Scientific Supplier No. DCE003CP
D (+) Cellobiose is a high-purity carbohydrate used in laboratory applications. This 25-gram quantity is ideal for experiments requiring a precise amount of cellobiose, a disaccharide composed of two glucose molecules. It is often used in biochemical research, especially in the study of enzymes, fermentation processes, and metabolic pathways involving cellobiose as a substrate. The product is of pharmaceutical grade, ensuring high quality and purity for accurate results in laboratory experiments. Its use extends to microbiology, molecular biology, and carbohydrate analysis protocols. Available in a convenient 25g bottle, perfect for routine testing and experiments.