Learn More
Medchemexpress LLC D-(+)-Cellobiose | 528-50-7 | MFCD00136034 | 25 MG
SDP
Catalog No. 5000422058
Encompass_Preferred
D-(+)-Cellobiose is a water-soluble disaccharide and a crucial intermediate in the degradation of cellulose and hemicellulose. It serves as a carbon source for microorganisms in fermentation processes, which convert it into ethanol or other metabolic byproducts. This makes D-(+)-Cellobiose particularly valuable for applications in biofuel production, such as bioethanol.
  • Water-soluble disaccharide
  • Important intermediate in cellulose and hemicellulose degradation
  • Utilized as a carbon source by microorganisms in fermentation
  • Converts into ethanol and other metabolic products
  • Significant application in biofuel production
  • For research use only
Catalog No. 50-004-22058 Supplier Medchemexpress LLC Supplier No. 25MGHYN2325R
D-(+)-Cellobiose is a water-soluble disaccharide and a crucial intermediate in the degradation of cellulose and hemicellulose. It serves as a carbon source for microorganisms in fermentation processes, which convert it into ethanol or other metabolic byproducts. This makes D-(+)-Cellobiose particularly valuable for applications in biofuel production, such as bioethanol.
  • Water-soluble disaccharide
  • Important intermediate in cellulose and hemicellulose degradation
  • Utilized as a carbon source by microorganisms in fermentation
  • Converts into ethanol and other metabolic products
  • Significant application in biofuel production
  • For research use only