Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(+)-cellobiose | 528-50-7 | MFCD00136034 | 99.8% | 342.30 | C12H22O11 | 10 MG
SDP

Catalog No. 5000289380
Encompass_Preferred

D-(+)-Cellobiose (Standard) is a high-purity analytical reference standard of the disaccharide D-(+)-cellobiose intended for research and analytical applications. It is supplied as a small packaged standard suitable for method development, calibration, and metabolite identification.

  • High purity (99.84%).
  • CAS number 528-50-7.
  • Molecular weight 342.30 g/mol.
  • White, water-soluble powder suitable for analytical use.
  • Intended for method development, calibration, and metabolite identification.

Catalog No. 50-002-89380 Supplier Medchemexpress LLC Supplier No. HYN2325R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(+)-Cellobiose (Standard) is a high-purity analytical reference standard of the disaccharide D-(+)-cellobiose intended for research and analytical applications. It is supplied as a small packaged standard suitable for method development, calibration, and metabolite identification.

  • High purity (99.84%).
  • CAS number 528-50-7.
  • Molecular weight 342.30 g/mol.
  • White, water-soluble powder suitable for analytical use.
  • Intended for method development, calibration, and metabolite identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.