Learn More
Medchemexpress LLC D-(+)-cellotetraose | 38819-01-1 | MFCD00079079 | 93.4% | 666.58 g/mol | C24H42O21 | 1 MG
SDP
Catalog No. 5000283007
Encompass_Preferred
D-(+)-Cellotetraose is a cellulose-derived tetrasaccharide supplied as a research reagent and analytical standard. It is a primary hydrolysis product of cellulose used in enzymatic studies, method development, and carbohydrate analysis. The material is provided in milligram quantities with reported purity of 93.37% and is characterized by formula C24H42O21 and molecular weight 666.58 g/mol.
  • Used as an analytical standard for HPLC and mass spectrometry.
  • Supports enzymatic hydrolysis and cellulase activity assays.
  • Provides a defined oligosaccharide for method development and calibration.
  • Reported purity 93.4% aiding reproducible experimental results.
  • Characterized by formula C24H42O21 and molecular weight 666.58 g/mol.
Catalog No. 50-002-83007 Supplier Medchemexpress LLC Supplier No. HYW1454981MG
D-(+)-Cellotetraose is a cellulose-derived tetrasaccharide supplied as a research reagent and analytical standard. It is a primary hydrolysis product of cellulose used in enzymatic studies, method development, and carbohydrate analysis. The material is provided in milligram quantities with reported purity of 93.37% and is characterized by formula C24H42O21 and molecular weight 666.58 g/mol.
  • Used as an analytical standard for HPLC and mass spectrometry.
  • Supports enzymatic hydrolysis and cellulase activity assays.
  • Provides a defined oligosaccharide for method development and calibration.
  • Reported purity 93.4% aiding reproducible experimental results.
  • Characterized by formula C24H42O21 and molecular weight 666.58 g/mol.