Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(+)-cellotriose | 33404-34-1 | MFCD00069842 | 98.0% | 504.44 g·mol⁻¹ | C18H32O16 | 5 MG
SDP

Catalog No. 5000283208
Encompass_Preferred

D-(+)-Cellotriose is a cello-oligosaccharide composed of three β-1,4-linked D-glucose units. It is formed during the enzymatic hydrolysis of cellulose and is used as a research reagent in studies of cellulolytic enzymes, carbohydrate metabolism, and glycobiology.

  • Oligosaccharide of three β-1,4-linked D-glucose units.
  • Intermediate in enzymatic hydrolysis of cellulose.
  • Molecular formula C18H32O16 and molecular weight 504.44 g·mol⁻¹.
  • CAS number 33404-34-1.
  • Purity 98.0% suitable for analytical and enzymatic assays.
  • Typical pack size 5 MG.
  • Supplied as a solid; handle per standard carbohydrate reagent procedures.

Catalog No. 50-002-83208 Supplier Medchemexpress LLC Supplier No. HYW145495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(+)-Cellotriose is a cello-oligosaccharide composed of three β-1,4-linked D-glucose units. It is formed during the enzymatic hydrolysis of cellulose and is used as a research reagent in studies of cellulolytic enzymes, carbohydrate metabolism, and glycobiology.

  • Oligosaccharide of three β-1,4-linked D-glucose units.
  • Intermediate in enzymatic hydrolysis of cellulose.
  • Molecular formula C18H32O16 and molecular weight 504.44 g·mol⁻¹.
  • CAS number 33404-34-1.
  • Purity 98.0% suitable for analytical and enzymatic assays.
  • Typical pack size 5 MG.
  • Supplied as a solid; handle per standard carbohydrate reagent procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.