Learn More
Cayman Chemical D-Celopentaose 10mg
SDP
Catalog No. 502913706
Encompass_Preferred
An oligosaccharide; has been used as a synthetic acceptor co-substrate to characterize the binding domains and enzyme kinetics of the E. coli pEtN transferase BcsG; has been used a substrate for a synthetic receptor designed to solubilize polysaccharides for use in tissue engineering and biological applications
Catalog No. 50-291-3706 Supplier Cayman Chemical Supplier No. 3530010 MG
An oligosaccharide; has been used as a synthetic acceptor co-substrate to characterize the binding domains and enzyme kinetics of the E. coli pEtN transferase BcsG; has been used a substrate for a synthetic receptor designed to solubilize polysaccharides for use in tissue engineering and biological applications