Learn More
Bachem b-Mercapto-b,b-cyclopentamethylene-propionyl-Tyr(Me)-Ile-Gln-Asn-Cys-Pro-Orn-Gly-NH2
SDP
Supplier:  Bachem 4034493.0001
Encompass_Preferred
(d(CH2)51,Tyr(Me)2,Orn8)-Oxytocin Trifluoroacetate, b-Mercapto-b,b-cyclopentamethylene-propionyl-Tyr(Me)-Ile-Gln-Asn-Cys-Pro-Orn-Gly-NH2, 77327-45-8, C48H74N12O12S2, Mr 1075.32, 1mg. [Pmp1, Tyr(OMe)2, Orn8] Vasotocin. Potent oxytocin antagonist with a pA2 value of 7.35 ± 0.08. (d(CH2)51,Tyr(Me)2,Orn8)-Vasotocin has been used to support the hypothesis that pituitary secretion of oxytocin is associated with coactivation of centrally projecting brain oxytocin pathways, some of which are causally related to the induced inhibition of food intake. Furthermore, this compound competitively inhibited arginine vasotocin (AVT) contractions without affecting AVT efficacy.
Catalog No. 50-259-900