Learn More
Bachem (d(CH2)51,Tyr(Me)2,Thr4,Orn8,Tyr-NH29)-Vasotocin Trifluoroacetate
SDP
Supplier:  Bachem 4015938.0001
Encompass_Preferred
(d(CH2)51,Tyr(Me)2,Thr4,Orn8,Tyr-NH29)-Vasotocin Trifluoroacetate, b-Mercapto-b,b-cyclopentamethylene-propionyl-Tyr(Me)-Ile-Thr-Asn-Cys-Pro-Orn-Tyr-NH2, 114056-26-7, C54H79N11O13S2, Mr 1154.42, 1mg. OVTA, Vasotocin analog. The oxytocin antagonist OVTA shows enhanced affinity for rat uterine oxytocin receptors after iodination.
Catalog No. 50-260-241