Learn More
Cayman Chemical D- CItramalIc AcIdlIthIu 10mg
SDP
Catalog No. 502984470
Encompass_Preferred
A biochemical intermediate involved in the anaerobic metabolism of glutamate through the methylaspartate pathway of C. tetanomorphum; also plays various roles in other aspects of bacterial metabolism
Catalog No. 50-298-4470 Supplier Cayman Chemical Supplier No. 1926010 MG
A biochemical intermediate involved in the anaerobic metabolism of glutamate through the methylaspartate pathway of C. tetanomorphum; also plays various roles in other aspects of bacterial metabolism