Learn More
Medchemexpress LLC D-Cl-amidine hydrochloride | 2985338-61-0 | 99.9% | 347.24 | C14H20Cl2N4O2 | 25 MG
SDP
Catalog No. 5000212236
Encompass_Preferred
D-Cl-amidine hydrochloride is the hydrochloride salt of a selective protein arginine deiminase 1 (PAD1) inhibitor for research use. It is supplied as a solid or as a DMSO solution and includes documented solubility and storage recommendations to support laboratory handling.
  • Selective PAD1 inhibitor with documented biological activity.
  • Hydrochloride salt form improves aqueous solubility.
  • High purity (99.91%) suitable for research applications.
  • Available as small solid quantities and as a 10 mM solution in DMSO.
  • Soluble in DMSO and water; sonication and warming recommended for dissolution.
  • Store protected from light under inert atmosphere at -20 °C; in solution, store at -80 °C for long-term stability.
Catalog No. 50-002-12236 Supplier Medchemexpress LLC Supplier No. HY100574D25MG
D-Cl-amidine hydrochloride is the hydrochloride salt of a selective protein arginine deiminase 1 (PAD1) inhibitor for research use. It is supplied as a solid or as a DMSO solution and includes documented solubility and storage recommendations to support laboratory handling.
  • Selective PAD1 inhibitor with documented biological activity.
  • Hydrochloride salt form improves aqueous solubility.
  • High purity (99.91%) suitable for research applications.
  • Available as small solid quantities and as a 10 mM solution in DMSO.
  • Soluble in DMSO and water; sonication and warming recommended for dissolution.
  • Store protected from light under inert atmosphere at -20 °C; in solution, store at -80 °C for long-term stability.