Learn More
Medchemexpress LLC D-Cl-amidine hydrochloride | 2985338-61-0 | 99.9% | 347.24 g/mol | C14H20Cl2N4O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100574D10MM 1ML
Encompass_Preferred
D-Cl-amidine hydrochloride is a potent, highly selective protein arginine deiminase 1 (PAD1) inhibitor supplied as a solid or as a ready-to-use 10 mM solution in DMSO. It is provided with high reported purity and includes recommended storage instructions for both solid and stock solution formats, making it suitable for biochemical assays and inhibitor studies.
  • Potent and highly selective PAD1 inhibition.
  • Available as solid or 10 mM solution in DMSO for workflow flexibility.
  • High reported purity (99.91%) for reproducible results.
  • Multiple pack sizes to accommodate small-scale and larger studies.
  • Recommended storage conditions for solid and stock solutions.
Catalog No. 50-002-12666