Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Cl-amidine hydrochloride | 2985338-61-0 | 99.9% | 347.24 g/mol | C14H20Cl2N4O2 | 100 MG
SDP

Catalog No. 5000212605
Encompass_Preferred

D-Cl-amidine hydrochloride is a research-grade small-molecule inhibitor provided as the hydrochloride salt and intended for in vitro and preclinical studies. It is supplied as a solid powder with high reported purity and should be handled and stored under recommended conditions to preserve stability.

  • Selective inhibitor of protein arginine deiminase 1 (PAD1).
  • Hydrochloride salt, solid form.
  • High purity (99.9%).
  • White to light yellow powder.
  • Store at -20°C, protect from light and moisture.
  • Available in research quantities such as 100 MG.

Catalog No. 50-002-12605 Supplier Medchemexpress LLC Supplier No. HY100574D100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Cl-amidine hydrochloride is a research-grade small-molecule inhibitor provided as the hydrochloride salt and intended for in vitro and preclinical studies. It is supplied as a solid powder with high reported purity and should be handled and stored under recommended conditions to preserve stability.

  • Selective inhibitor of protein arginine deiminase 1 (PAD1).
  • Hydrochloride salt, solid form.
  • High purity (99.9%).
  • White to light yellow powder.
  • Store at -20°C, protect from light and moisture.
  • Available in research quantities such as 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.