Learn More
Medchemexpress LLC D-Cl-amidine hydrochloride | 2985338-61-0 | 99.91% | 347.24 | 5 MG
SDP
Catalog No. 5000365973
Encompass_Preferred
D-Cl-amidine hydrochloride is a potent and highly selective PAD1 inhibitor. It is well-tolerated with no significant toxicity and is suitable for research use.
  • Significantly decreases cell viability in MDA-MB-231 cells
  • Increases caspase 3 activity, indicating increased apoptosis
  • Detectable in serum after administration
Catalog No. 50-003-65973 Supplier Medchemexpress LLC Supplier No. 5MGHY100574D
Add to Cart
Add to Cart
D-Cl-amidine hydrochloride is a potent and highly selective PAD1 inhibitor. It is well-tolerated with no significant toxicity and is suitable for research use.
  • Significantly decreases cell viability in MDA-MB-231 cells
  • Increases caspase 3 activity, indicating increased apoptosis
  • Detectable in serum after administration