Learn More
Medchemexpress LLC D-Cl-amidine hydrochloride | 2985338-61-0 | MFCD33398967 | 99.9% | 347.24 g/mol | C14H20Cl2N4O2 | 10 MG
SDP
Catalog No. 5000212931
Encompass_Preferred
D-Cl-amidine hydrochloride is a potent, selective inhibitor of protein arginine deiminase 1 (PAD1). Supplied as the hydrochloride salt and available as a solid or as a 10 mM DMSO solution, it is used in biochemical and cell-based studies to probe PAD1-mediated citrullination, influence on cell viability, and induction of apoptosis.
  • High purity (≈99.9%) suitable for research use.
  • Available as small solid batches and a DMSO solution for flexible dosing.
  • Readily soluble in DMSO and water for assay preparation.
  • Documented in vitro activity and measurable exposure in vivo.
  • Suitable for biochemical and cell-based assays.
  • Store protected from light and under inert atmosphere for stability.
Catalog No. 50-002-12931 Supplier Medchemexpress LLC Supplier No. HY100574D10MG
D-Cl-amidine hydrochloride is a potent, selective inhibitor of protein arginine deiminase 1 (PAD1). Supplied as the hydrochloride salt and available as a solid or as a 10 mM DMSO solution, it is used in biochemical and cell-based studies to probe PAD1-mediated citrullination, influence on cell viability, and induction of apoptosis.
  • High purity (≈99.9%) suitable for research use.
  • Available as small solid batches and a DMSO solution for flexible dosing.
  • Readily soluble in DMSO and water for assay preparation.
  • Documented in vitro activity and measurable exposure in vivo.
  • Suitable for biochemical and cell-based assays.
  • Store protected from light and under inert atmosphere for stability.