Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Cl-amidine hydrochloride | 2985338-61-0 | MFCD33398967 | 99.9% | 347.24 g·mol⁻¹ | C14H20Cl2N4O2 | 50 MG
SDP

Catalog No. 5000211566
Encompass_Preferred

D-Cl-amidine hydrochloride is the hydrochloride salt of D-Cl-amidine, a potent and selective inhibitor of protein arginine deiminase 1 (PAD1) used in biochemical and cellular research. It is supplied as a high-purity reagent in powder and DMSO solution formats for enzymatic assays and cellular studies.

  • High purity suitable for research use.
  • Selective inhibition of protein arginine deiminase 1 (PAD1) for target-specific studies.
  • Available in multiple sizes and a ready-to-use DMSO solution for convenience.
  • Molecular formula C14H20Cl2N4O2 and molecular weight 347.24 g·mol⁻¹.
  • Supplied with product datasheet and quality information for reproducibility.

Catalog No. 50-002-11566 Supplier Medchemexpress LLC Supplier No. HY100574D50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Cl-amidine hydrochloride is the hydrochloride salt of D-Cl-amidine, a potent and selective inhibitor of protein arginine deiminase 1 (PAD1) used in biochemical and cellular research. It is supplied as a high-purity reagent in powder and DMSO solution formats for enzymatic assays and cellular studies.

  • High purity suitable for research use.
  • Selective inhibition of protein arginine deiminase 1 (PAD1) for target-specific studies.
  • Available in multiple sizes and a ready-to-use DMSO solution for convenience.
  • Molecular formula C14H20Cl2N4O2 and molecular weight 347.24 g·mol⁻¹.
  • Supplied with product datasheet and quality information for reproducibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.