Learn More
Research Products International Corp D-Cycloserine, 1 Gram
SDP
Catalog No. 50213262
Encompass_Preferred
D-cycloserine is an antibiotic and structural analog of the amino acid D-alanine.  It inhibits cell wall synthesis in certain bacteria, including Mycobacterium tuberculosis.
While it was initially identified as an antibiotic, D-cycloserine has gained attention for its activity as a partial agonist at the N-methyl-D-aspartate (NMDA) receptor, a type of glutamate receptor in the brain. Researchers study the mechanisms of neurotransmission and synaptic plasticity due to its interactions with NMDA receptors. 
Catalog No. 50-213-262 Supplier Research Products International Corp Supplier No. C810001.0
D-cycloserine is an antibiotic and structural analog of the amino acid D-alanine.  It inhibits cell wall synthesis in certain bacteria, including Mycobacterium tuberculosis.
While it was initially identified as an antibiotic, D-cycloserine has gained attention for its activity as a partial agonist at the N-methyl-D-aspartate (NMDA) receptor, a type of glutamate receptor in the brain. Researchers study the mechanisms of neurotransmission and synaptic plasticity due to its interactions with NMDA receptors. 