Learn More
Apexbio Technology LLC D-Cycloserine 68-41-7 500mg
SDP
Catalog No. 5000094708
Encompass_Preferred
D-Cycloserine (CAS 68-41-7) is a small-molecule inhibitor targeting bacterial cell wall peptidoglycan biosynthesis enzymes It is designed to inhibit key enzymes involved in peptidoglycan formation thereby interfering with bacterial cell wall synthesis D-Cycloserine exerts its biological activity primarily through blocking the enzymes essential for bacterial cell wall construction It demonstrates inhibitory activity with reported IC50 values in the micromolar range against cell wall synthesis enzymes depending on experimental conditions Based on these pharmacological properties D-Cycloserine holds research potential in studies on multidrug-resistant tuberculosis antibacterial resistance mechanisms and as a partial agonist of the NMDA receptor for neuroscientific exploration of cognitive-related neuronal activity
Catalog No. 50-000-94708 Supplier Apexbio Technology LLC Supplier No. B1721500
D-Cycloserine (CAS 68-41-7) is a small-molecule inhibitor targeting bacterial cell wall peptidoglycan biosynthesis enzymes It is designed to inhibit key enzymes involved in peptidoglycan formation thereby interfering with bacterial cell wall synthesis D-Cycloserine exerts its biological activity primarily through blocking the enzymes essential for bacterial cell wall construction It demonstrates inhibitory activity with reported IC50 values in the micromolar range against cell wall synthesis enzymes depending on experimental conditions Based on these pharmacological properties D-Cycloserine holds research potential in studies on multidrug-resistant tuberculosis antibacterial resistance mechanisms and as a partial agonist of the NMDA receptor for neuroscientific exploration of cognitive-related neuronal activity