Learn More
Bioworld D-Cycloserine, 1 g
Supplier:  Bioworld 403001321
Encompass_Preferred
Antibiotic and a partial agonist of NMDA receptors at the glycine modulatory site. Interferes with bacterial cell wall synthesis by competing with D-Alanine for incorporation into the cell wall. Active against most mycobacteria with some activity against gram negative bacteria.Appearance: White to pale yellow crystalline PowderDensity : 1.278 g/cm3Melting Point: 147 °CRTECS Number: NY2975000
Catalog No. 50-255-685