Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Cycloserine, G-Biosciences
SCI_ED

Catalog No. S692101
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S692101 5 g
S100489 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S692101 Supplier G-Biosciences Supplier No. RC991
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Cycloserine is an amino acid analog antibiotic effective against a wide variety of bacteria. It is commonly used against Mycobacterium tuberculosis.

It is suitable for use as a selection agent in a variety of isolation methods. D-cycloserine inhibits a variety of enzymes, is a partial NMDA receptor agonist, and may improve memory and cognition.

Chemical Identifiers

CAS 68-41-7
Molecular Weight (g/mol) 102.09 g/mol

Specifications

Color White
Quantity 5 g
Percent Purity >98%
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material D-Cycloserine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.