Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-cysteine hydrochloride hydrate | 207121-46-8 | MFCD00150051 | 97.0% | 175.63 g/mol | C3H10ClNO3S | 500 G
SDP

Catalog No. 5000210474
Encompass_Preferred

D-cysteine hydrochloride hydrate is the hydrochloride hydrate salt of the D-enantiomer of cysteine, supplied as an amino acid derivative for research and synthetic chemistry applications. It is a white to off-white solid used as a building block in biochemical studies and small-molecule synthesis.

  • purity: 97.0% (as provided by supplier)
  • molecular weight: 175.63 g/mol
  • chemical formula: C3H10ClNO3S
  • cas number: 207121-46-8
  • appearance: white to off-white solid
  • storage: store sealed at 4°C and protect from moisture; in solution, store at -80°C for long-term and -20°C for short-term
  • packaging: available in sample, 100 g, 500 g, and larger quantities
  • typical uses: used as a reagent or building block in biochemical research and synthetic chemistry

Catalog No. 50-002-10474 Supplier Medchemexpress LLC Supplier No. HYW055811500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-cysteine hydrochloride hydrate is the hydrochloride hydrate salt of the D-enantiomer of cysteine, supplied as an amino acid derivative for research and synthetic chemistry applications. It is a white to off-white solid used as a building block in biochemical studies and small-molecule synthesis.

  • purity: 97.0% (as provided by supplier)
  • molecular weight: 175.63 g/mol
  • chemical formula: C3H10ClNO3S
  • cas number: 207121-46-8
  • appearance: white to off-white solid
  • storage: store sealed at 4°C and protect from moisture; in solution, store at -80°C for long-term and -20°C for short-term
  • packaging: available in sample, 100 g, 500 g, and larger quantities
  • typical uses: used as a reagent or building block in biochemical research and synthetic chemistry

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.