Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Cysteine hydrochloride monohydrate, 99%

Catalog No. AAH2710706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2710706 5 g
AAH2710703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2710706 Supplier Thermo Scientific Chemicals Supplier No. H2710706
  • $376.00
    $303.65 / Each of 1

    Save $72.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 207121-46-8 | C3H10ClNO3S | 175.627 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207121-46-8
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.627
MDL Number MFCD00150051
InChI Key QIJRTFXNRTXDIP-YBBRRFGFSA-N
Synonym d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
PubChem CID 10176341
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Melting Point 184°C to 186°C
Quantity 5 g
Formula Weight 175.64 (157.62 Anhydrous)
Optical Rotation −5.5° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-Cysteine hydrochloride monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 251-043-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.