Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Cysteine Hydrochloride Monohydrate 99.0+%, TCI America™
SDP

Catalog No. C13295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13295G 5 g
C132925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13295G Supplier TCI America Supplier No. C13295G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 207121-46-8
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.627
MDL Number MFCD00150051
InChI Key QIJRTFXNRTXDIP-YBBRRFGFSA-N
Synonym d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
PubChem CID 10176341
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 157.61
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Cysteine Hydrochloride Monohydrate
EINECSNumber (2)-3245
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.