Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Cysteine Methyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C21745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21745G 5 g
C217425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21745G Supplier TCI America Supplier No. C21745G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 70361-61-4
Molecular Formula C4H10ClNO2S
Molecular Weight (g/mol) 171.639
MDL Number MFCD00173915
InChI Key WHOHXJZQBJXAKL-AENDTGMFSA-N
Synonym H-D-Cys-OMe.HCl
PubChem CID 637545
IUPAC Name methyl (2S)-2-amino-3-sulfanylpropanoate;hydrochloride
SMILES COC(=O)C(CS)N.Cl

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 5 g
Formula Weight 171.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Cysteine Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.