Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Cystine, 98%

Catalog No. AAL1377206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1377206 5 g
AAL1377203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1377206 Supplier Thermo Scientific Chemicals Supplier No. L1377206
Add to Cart

CAS: 349-46-2 | C6H12N2O4S2 | 240.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-46-2
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.29
MDL Number MFCD00002610
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym d-cystine, 2-amino-3-2s-2-amino-2-carboxyethyl disulfanyl propanoic acid
PubChem CID 6857538
ChEBI CHEBI:35494
IUPAC Name (2S)-2-amino-3-[[(2S)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

Specifications

Melting Point ∽250°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 1728093
Merck Index 14,2782
Optical Rotation +219° (c=1 in 1N HCl)
Formula Weight 240.3
Percent Purity 98%
Chemical Name or Material D-Cystine
Hazard Category H302-H315-H320-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 206-486-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.