Learn More
Cayman Chemical D-DesthIobIotIn 250mg
SDP
Catalog No. 502990829
Encompass_Preferred
A protein cross-linking agent and biotin synthesis precursor; has been used in the preparation of agarose matrices for affinity-based isolation of streptavidin-fluorophore conjugates
Catalog No. 50-299-0829 Supplier Cayman Chemical Supplier No. 30418250 MG
A protein cross-linking agent and biotin synthesis precursor; has been used in the preparation of agarose matrices for affinity-based isolation of streptavidin-fluorophore conjugates