Learn More
Cayman Chemical D-Desthiobiotin, Purity: ≥95%, CAS: 533-48-2, 500 mg
SDP
Catalog No. 502992035
Encompass_Preferred
A protein cross-linking agent and biotin synthesis precursor; has been used in the preparation of agarose matrices for affinity-based isolation of streptavidin-fluorophore conjugates
Catalog No. 50-299-2035 Supplier Cayman Chemical Supplier No. 30418500 MG
A protein cross-linking agent and biotin synthesis precursor; has been used in the preparation of agarose matrices for affinity-based isolation of streptavidin-fluorophore conjugates