Learn More
Apexbio Technology LLC D-Desthiobiotin 533-48-2 100mg
SDP
Catalog No. 5000090141
Encompass_Preferred
D-Desthiobiotin (CAS 533-48-2) is a derivative of biotin lacking the thiophene ring It binds to avidin and streptavidin with lower affinity compared to biotin enabling reversible interaction in affinity purification and protein chromatography protocols This property facilitates gentle elution of biotinylated proteins or other biomolecules without harsh conditions D-Desthiobiotin is widely employed for the labeling detection and isolation of proteins and cells in biochemical and cell-based assays providing an efficient tool for applications requiring temporary and specific capture of biomolecular targets
Catalog No. 50-000-90141 Supplier Apexbio Technology LLC Supplier No. A8014100
D-Desthiobiotin (CAS 533-48-2) is a derivative of biotin lacking the thiophene ring It binds to avidin and streptavidin with lower affinity compared to biotin enabling reversible interaction in affinity purification and protein chromatography protocols This property facilitates gentle elution of biotinylated proteins or other biomolecules without harsh conditions D-Desthiobiotin is widely employed for the labeling detection and isolation of proteins and cells in biochemical and cell-based assays providing an efficient tool for applications requiring temporary and specific capture of biomolecular targets