Learn More
Medchemexpress LLC D-Desthiobiotin | 533-48-2 | 99.7% | 250 MG
SDP
Catalog No. 5000341473
Encompass_Preferred
D-Desthiobiotin is a biotin derivative utilized in affinity and protein chromatography. It is also suitable for the labeling, detection, and isolation of proteins and cells, making it valuable for various research and analytical applications.
  • Biotin derivative
  • Used in affinity chromatography
  • Used in protein chromatography
  • Suitable for protein labeling
  • Suitable for cell labeling
  • Aids in detection of proteins and cells
  • Aids in isolation of proteins and cells
  • Intended for research and analytical applications
Catalog No. 50-003-41473 Supplier Medchemexpress LLC Supplier No. 250MGHY128699
D-Desthiobiotin is a biotin derivative utilized in affinity and protein chromatography. It is also suitable for the labeling, detection, and isolation of proteins and cells, making it valuable for various research and analytical applications.
  • Biotin derivative
  • Used in affinity chromatography
  • Used in protein chromatography
  • Suitable for protein labeling
  • Suitable for cell labeling
  • Aids in detection of proteins and cells
  • Aids in isolation of proteins and cells
  • Intended for research and analytical applications