Learn More
IBA LifeSciences Desthiobiotin; 5 g
Catalog No. NC0395527
Encompass_Preferred
Desthiobiotin is a modified form of biotin that displaces Strep-tagII and Twin-Strep-tag fusion proteins from the modified biotin-binding pocket of Strep-Tactin in a competitive manner, resulting in a mild elution. Desthiobiotin powder can be used for preparation of Strep-Tactin elution buffer, Buffer E. Please note that desthiobiotin is not applicable for elution from Strep-TactinXT resins.
Catalog No. NC0395527 Supplier IBA LifeSciences Supplier No. 21000005
Desthiobiotin is a modified form of biotin that displaces Strep-tagII and Twin-Strep-tag fusion proteins from the modified biotin-binding pocket of Strep-Tactin in a competitive manner, resulting in a mild elution. Desthiobiotin powder can be used for preparation of Strep-Tactin elution buffer, Buffer E. Please note that desthiobiotin is not applicable for elution from Strep-TactinXT resins.