Learn More
Medchemexpress LLC HY-128699 5mg Medchemexpress, D-Desthiobiotin CAS:533-48-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1286995MG
Encompass_Preferred
Medchemexpress, HY-128699 5mg D-Desthiobiotin CAS:533-48-2 D-Desthiobiotin is a biotin derivative used in affinity chromatography and protein chromatography, also can be used for protein and cell labeling, detection and isolation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2629