Learn More
Sigma Aldrich Fine Chemicals Biosciences d-Desthiobiotin, 533-48-2, MFCD00213804, 1g
Catalog No. NC1606884
Encompass
Empirical Formula (Hill Notation) C10H18N2O3, ≥98% (TLC), Molecular Weight 214.26, Synonym: 5-Methyl-2-oxo-4-imidazolidinehexanoic acid. d-Desthiobiotin is used in affinity chromatography and protein chromatography. d-Desthiobiotin can be used for protein labeling, detection and isolation.
Catalog No. NC1606884 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D14111G
Empirical Formula (Hill Notation) C10H18N2O3, ≥98% (TLC), Molecular Weight 214.26, Synonym: 5-Methyl-2-oxo-4-imidazolidinehexanoic acid. d-Desthiobiotin is used in affinity chromatography and protein chromatography. d-Desthiobiotin can be used for protein labeling, detection and isolation.