Learn More
Medchemexpress LLC D-Desthiobiotin (Standard) | 533-48-2 | 1 G
SDP
Catalog No. 5000341028
Encompass_Preferred
D-Desthiobiotin is a biotin derivative that increases testosterone. It is utilized in affinity and protein chromatography, and can also be employed for protein and cell labeling, detection, and isolation.
  • Biotin derivative
  • Increases testosterone
  • Used in affinity chromatography
  • Used in protein chromatography
  • Employed for protein and cell labeling
  • Employed for protein and cell detection
  • Employed for protein and cell isolation
Catalog No. 50-003-41028 Supplier Medchemexpress LLC Supplier No. HY1286991G
D-Desthiobiotin is a biotin derivative that increases testosterone. It is utilized in affinity and protein chromatography, and can also be employed for protein and cell labeling, detection, and isolation.
  • Biotin derivative
  • Increases testosterone
  • Used in affinity chromatography
  • Used in protein chromatography
  • Employed for protein and cell labeling
  • Employed for protein and cell detection
  • Employed for protein and cell isolation