Learn More
Crystal Chem Inc D-Dimer Assay Kit, 96-test
SDP
Catalog No. 501947685
Encompass
The D-Dimer Assay is used to quantify levels of human D-Dimer in plasma using only a 3 µL sample volume.Highlights: Highly sensitive immunoturbidimetric assay. Dynamic range of 0.23 to 8.0 mg/mL FEU. Designed to work with most chemistry analyzers
Catalog No. 50-194-7685 Supplier Crystal Chem Inc Supplier No. 80279
The D-Dimer Assay is used to quantify levels of human D-Dimer in plasma using only a 3 µL sample volume.Highlights: Highly sensitive immunoturbidimetric assay. Dynamic range of 0.23 to 8.0 mg/mL FEU. Designed to work with most chemistry analyzers