Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories D-Dimer Assay

Catalog No. 23700617
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Diagnostic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-700-617 Diagnostic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-700-617 Supplier Diazyme Laboratories Supplier No. DZ179AK
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diazyme D-Dimer Assay is for the quantitative determination of fibrinogen/fibrin degradation products (D-Dimer) in human plasma.

  • Highly sensitive liquid stable latex enhanced immunoturbidimetric test kit, designed to work on most open clinical chemistry analyzers to provide rapid results with reduced reagent cost
  • The D-Dimer Assay has a linearity of 0.15 - 8.0 μg/mL FEU with a correlation coefficient value of 0.939, slope of 0.979 and y intercept of -0.106
  • The following substances do not interfere with this assay at the levels tested less than 10% bias: Hemoglobin up to 500 mg/dL, Bilirubin 40 mg/dL, Bilirubin conjugated 40mg/dL, Triglycerides 1000 mg/dL, Ascorbic Acid 176 mg/dL, Rheumatoid Factor 100 IU/mL, Heparin 1.5 IU/mL and HAMA 490 ng/mL
  • Measurement of D-Dimer is used as an aid in detecting the presence of intravascular coagulation and fibrinolysis
  • For in vitro diagnostic use only

Specifications

Certifications/Compliance 510(k) Cleared, EU: CE IVD
Content And Storage D-Dimer assay reagents, calibrators, and controls should be stored at 2 to 8°C. DO NOT FREEZE
Description D-Dimer Assay Kit
CE Marker Yes
Concentration Lab dependent
Type Diagnostic
Includes R1 and R2
Linearity Eleven levels of the D-Dimer linearity set were prepared by diluting a specimen containing 8.0 μg/mL FEU with saline according to Clinical and Laboratory Standards Institute EP6-A and tested on Modular P. Diazyme D-Dimer assay is linear from 0.15 to 8.0 μg/mL FEU.
Packaging Type Universal
Stability The reagents, calibrators, and controls are sta-ble when stored as instructed until the expiration date on the label. Open reagent bottles are stable for 4 weeks if stored on board the refrigerated compartment of the Roche Modular P analyzer.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.