Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories D-Dimer Control

Catalog No. 23700619
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Diagnostic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-700-619 Diagnostic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-700-619 Supplier Diazyme Laboratories Supplier No. DZ179ACON
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diazyme D-Dimer Control set is for the testing of D-Dimer in human plasma.

  • Highly sensitive liquid stable latex enhanced immunoturbidimetric test kit, designed to work on most open clinical chemistry analyzers to provide rapid results with reduced reagent cost
  • The D-Dimer Assay has a linearity of 0.15 - 8.0 μg/mL FEU with a correlation coefficient value of 0.939, slope of 0.979 and y intercept of -0.106
  • The following substances do not interfere with this assay at the levels tested less than 10% bias: Hemoglobin up to 500 mg/dL, Bilirubin 40 mg/dL, Bilirubin conjugated 40mg/dL, Triglycerides 1000 mg/dL, Ascorbic Acid 176 mg/dL, Rheumatoid Factor 100 IU/mL, Heparin 1.5 IU/mL and HAMA 490 ng/mL
  • Measurement of D-Dimer is used as an aid in detecting the presence of intravascular coagulation and fibrinolysis
  • For in vitro diagnostic use only

Specifications

Certifications/Compliance 510(k) Cleared, EU: CE IVD
Content And Storage 2°C to 8°C. DO NOT FREEZE. Note: Store CONTROL tightly capped when not in use.
Description D-Dimer Assay Control Set
CE Marker Yes
Concentration Lab dependent
Format 2 x 1 mL Vial
Type Diagnostic
Includes The Diazyme D-Dimer Control Set is a two-level set that is supplied in lyophilized form (2 x 1 mL). The controls are manufactured from human serum. The concentrations of the controls are lot specific and D-Dimer concentrations are expressed in μg/mL FEU.
Stability Unopened vials: stability is up to the expiration date on label when stored at 2 to 8°C After reconstitution: stability is 14 days when stored at 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.