Learn More
BioVendor, LLC D-Dimer Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL013R
Encompass
D-dimer, the final degradation product of cross-linked fibrin, is typically elevated in patients with acute venous thromboembolism. D-dimer testing is an integral part of validated algorithms for the diagnosis of deep-vein thrombosis (DVT) and pulmonary embolism (PE). Type: Sandwich ELISA. Sample type: serum, plasma. Sample requirement: 40 ¿l/well. Calibration range: 5-160ng/ml. Limit of detection: 1.25ng/ml. Other names: Fragment D-dimer, Fibrin degradation fragment, DVT - D-dimer, PE - D-dimer.
Catalog No. 50-240-6353