Learn More
MOLECULAR DEPOT LLC D-Dimer mAb - 0.5 mg
Catalog No. 5000013383
Encompass_Preferred
D-Dimer Monoclonal Antibody (0 5 mg) is an affinity-purified mouse monoclonal antibody specific to human D-dimer Tested in several immunoassay platforms including ELISA CLIA Immunoturbidimetry and more tested matching clones are available Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only
Catalog No. 50-000-13383 Supplier MOLECULAR DEPOT LLC Supplier No. B2010244
D-Dimer Monoclonal Antibody (0 5 mg) is an affinity-purified mouse monoclonal antibody specific to human D-dimer Tested in several immunoassay platforms including ELISA CLIA Immunoturbidimetry and more tested matching clones are available Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only