Learn More
General Laboratory Products D/E Neutralizing Broth, (DE Neutralizing Broth), 99 mL, 12/cs
Catalog No. NC0739013
Encompass_Preferred
D/E Neutralizing Broth - 99ml, 8 oz. wide mouth glass bottle, 12 per case. For testing and neutralizing of antiseptics and disinfectants.
Catalog No. NC0739013 Supplier General Laboratory Products Supplier No. 8WM120099
D/E Neutralizing Broth - 99ml, 8 oz. wide mouth glass bottle, 12 per case. For testing and neutralizing of antiseptics and disinfectants.