Learn More
Nelson Jameson Company D/E BROTH SPONGE-STICK 10/ML
Catalog No. NC1055807
Encompass
This item has a minimum order quantity of 2 per supplier requirements. •Stick-mounted sponge design allows the ability to access hard to reach areas without directly handling the sponge. •Stick-mounted sponge design provides the ability to easily snap off the sponge from the handle after swabbing. •Simplified packaging minimizes handling and preparation and reduces waste. •1.5 x 3 inch biocide-free sponge does not affect organism viability. 100/pk
Catalog No. NC1055807 Supplier Nelson Jameson Company Supplier No. 3513065 CY
This item has a minimum order quantity of 2 per supplier requirements. •Stick-mounted sponge design allows the ability to access hard to reach areas without directly handling the sponge. •Stick-mounted sponge design provides the ability to easily snap off the sponge from the handle after swabbing. •Simplified packaging minimizes handling and preparation and reduces waste. •1.5 x 3 inch biocide-free sponge does not affect organism viability. 100/pk