Learn More
NEOGEN D/E Neutralizing Agar with Tween, 500 g
SDP
Catalog No. 502015177
Encompass_Preferred
D/E Neutralizing Agar with Tween is used for the isolation of microorganisms from sanitized environmental surfaces and is not intended for use in the diagnosis of disease or other conditions in humans.D/E Neutralizing Agar with Tween was developed by Dey and Engley to neutralize a broad spectrum of disinfectants and preservative antimicrobial chemicals, including quaternary ammonium compounds, phenolics, iodine, chlorine preparations, mercurials, formaldehyde, and glutaraldehyde. D/E Neutralizing media neutralize higher concentrations of residual antimicrobials when compared with other standard neutralizing formulations, such as Letheen media, Thioglycolate media, and Neutralizing Buffer.For more information visit https://www.neogen.com/solutions/Microbiology/de-neutralizing-agar-with-tween
Catalog No. 50-201-5177 Supplier NEOGEN Supplier No. 700002983
D/E Neutralizing Agar with Tween is used for the isolation of microorganisms from sanitized environmental surfaces and is not intended for use in the diagnosis of disease or other conditions in humans.D/E Neutralizing Agar with Tween was developed by Dey and Engley to neutralize a broad spectrum of disinfectants and preservative antimicrobial chemicals, including quaternary ammonium compounds, phenolics, iodine, chlorine preparations, mercurials, formaldehyde, and glutaraldehyde. D/E Neutralizing media neutralize higher concentrations of residual antimicrobials when compared with other standard neutralizing formulations, such as Letheen media, Thioglycolate media, and Neutralizing Buffer.For more information visit https://www.neogen.com/solutions/Microbiology/de-neutralizing-agar-with-tween