Learn More
NEOGEN D/E Neutralizing Broth without Tween, 5 kg
SDP
Catalog No. 502015502
Encompass_Preferred
D/E Neutralizing Broth is used to neutralize and determine the bactericidal activity of antiseptics and disinfectants. D/E Neutralizing Broth without Tween is not intended for use in the diagnosis of disease or other conditions in humans.D/E Neutralizing Broth was developed by Dey and Engley to neutralize a broad spectrum of disinfectants and preservative antimicrobial chemicals, including quaternary ammonium compounds, phenolics, iodine, chlorine preparations, mercurials, formaldehyde, and glutaraldehyde. D/E Neutralizing media neutralize higher concentrations of residual antimicrobials when compared with other standard neutralizing formulations, such as Letheen media, Thioglycollate media, and Neutralizing Buffer.For more information visit https://www.neogen.com/solutions/Microbiology/de-neutralizing-broth-without-tween
Catalog No. 50-201-5502 Supplier NEOGEN Supplier No. 700003317
D/E Neutralizing Broth is used to neutralize and determine the bactericidal activity of antiseptics and disinfectants. D/E Neutralizing Broth without Tween is not intended for use in the diagnosis of disease or other conditions in humans.D/E Neutralizing Broth was developed by Dey and Engley to neutralize a broad spectrum of disinfectants and preservative antimicrobial chemicals, including quaternary ammonium compounds, phenolics, iodine, chlorine preparations, mercurials, formaldehyde, and glutaraldehyde. D/E Neutralizing media neutralize higher concentrations of residual antimicrobials when compared with other standard neutralizing formulations, such as Letheen media, Thioglycollate media, and Neutralizing Buffer.For more information visit https://www.neogen.com/solutions/Microbiology/de-neutralizing-broth-without-tween