Learn More
General Laboratory Products D/E Neutralizing Broth, (DE Neutralizing Broth), 9 mL, 100/cs
Catalog No. NC0504802
Encompass_Preferred
D/E Neutralizing Broth - 9ml, 100 per case. For testing and neutralizing of antiseptics and disinfectants.
Catalog No. NC0504802 Supplier General Laboratory Products Supplier No. 20T12009
D/E Neutralizing Broth - 9ml, 100 per case. For testing and neutralizing of antiseptics and disinfectants.