Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD D/E Neutralizing Broth Ready-to use- Bottled Media

Catalog No. 22349721 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Fluid D; Quantity: 10 x 90mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-349-721 Fluid D; Quantity: 10 x 90mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-349-721 Supplier BD Supplier No. 257450

Used for determining bactericidal activity of antiseptics and disinfectants based on neutralizing chemical and detecting organisms remaining after treatment

TRUSTED_SUSTAINABILITY

Specifications

Format Ready-to-Use Bottles Media
Quantity 10 x 90 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.