Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D.E.R.™ BisphenolA diglycidyl ether, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013319 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3319 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3319 Supplier MilliporeSigma Supelco Supplier No. 15138500MGF
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 1675-54-3
Product Line D.E.R.
Physical Form Neat
Linear Formula C21H24O4
MDL Number MFCD00080480
Melting Point 40°C to 44°C
Synonym 2,2-Bis[4-(glycidyloxy)phenyl]propane; 4,4′-Isopropylidenediphenol diglycidyl ether; BADGE; D.E.R.(TM) 332
Type Analytical Standard
Quantity 500 mg
Formula Weight 340.41
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less

D.E.R. is a trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.