Learn More
Crescent Chemical Co Inc D.E.R. 736 PRACT.
SDP
Catalog No. 50247472
Encompass_Preferred
D.E.R. 736 pract. Shorter chain than D.E.R. 732, lower viscosity, gives less flexible blocks. Epoxy equivalent weight 175 - 205; viscosity 0.03 - 0.06 Pas at 25 C.Used in electron microscopy. Size - 100ML Storage Conditions - +15 C TO +30 C
Catalog No. 50-247-472 Supplier Crescent Chemical Co Inc Supplier No. SE18247.01
D.E.R. 736 pract. Shorter chain than D.E.R. 732, lower viscosity, gives less flexible blocks. Epoxy equivalent weight 175 - 205; viscosity 0.03 - 0.06 Pas at 25 C.Used in electron microscopy. Size - 100ML Storage Conditions - +15 C TO +30 C