Learn More
Medchemexpress LLC D-erythro-dihydrosphingosine | 764-22-7 | MFCD00079141 | 98.0% | 301.51 | C18H39NO2 | 5 MG
SDP
Catalog No. 5000300287
Encompass_Preferred
D-erythro-dihydrosphingosine is a sphingolipid metabolite provided as an analytical research standard. It acts as a direct inhibitor of cytosolic phospholipase A2α (cPLA2α) and is intended for use in biochemical and cell-based studies of sphingolipid metabolism and signaling. The compound is supplied as a white to off-white solid with reported high purity.
Catalog No. 50-003-00287 Supplier Medchemexpress LLC Supplier No. HYW0198385MG
D-erythro-dihydrosphingosine is a sphingolipid metabolite provided as an analytical research standard. It acts as a direct inhibitor of cytosolic phospholipase A2α (cPLA2α) and is intended for use in biochemical and cell-based studies of sphingolipid metabolism and signaling. The compound is supplied as a white to off-white solid with reported high purity.